Product Details

SNP ID
rs1367950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3715004 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATAGCCATCTGTTTCAGAAAAC[C/T]GGCATTCTGGCTGTTGCCTAAATAG
Phenotype
MIM: 609240 MIM: 604682
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSG2 PubMed Links

Gene Details

Gene
GSG2
Gene Name
germ cell associated 2, haspin
There are no transcripts associated with this gene.

Gene
ITGAE
Gene Name
integrin subunit alpha E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002208.4 Intron NP_002199.3
XM_011523823.2 Intron XP_011522125.1
XM_011523825.2 Intron XP_011522127.1
XM_011523827.2 Intron XP_011522129.1
XM_011523828.2 Intron XP_011522130.1
XM_017024586.1 Intron XP_016880075.1
XM_017024587.1 Intron XP_016880076.1

View Full Product Details