Product Details

SNP ID
rs5197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153905611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACTGGACACCCGGGACCCGCT[A/G]CTAGCCCGGGCGGAGCTGGCGCTGC
Phenotype
MIM: 300023 MIM: 300538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP4 PubMed Links

Gene Details

Gene
ARHGAP4
Gene Name
Rho GTPase activating protein 4
There are no transcripts associated with this gene.

Gene
AVPR2
Gene Name
arginine vasopressin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000054.4 176 Silent Mutation CTA,CTG L35L NP_000045.1
NM_001146151.1 176 Silent Mutation CTA,CTG L35L NP_001139623.1
XM_006724828.3 176 Silent Mutation CTA,CTG L35L XP_006724891.1

View Full Product Details