Product Details

SNP ID
rs4325604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3215714 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGATTTCTTCTACATCCTCTTC[C/T]TGTTCATTTACCCCATCACATTGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1A1 PubMed Links

Gene Details

Gene
OR1A1
Gene Name
olfactory receptor family 1 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014565.2 94 Silent Mutation CTG,TTG L32L NP_055380.2

View Full Product Details