Product Details

SNP ID
rs1144846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120710667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCACGGAGCAGGAGCTGCTGGCG[A/C]TGGACACCATCCGGCCCGAGCACGT
Phenotype
MIM: 613802
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MLEC PubMed Links

Gene Details

Gene
MLEC
Gene Name
malectin
There are no transcripts associated with this gene.

Gene
UNC119B
Gene Name
unc-119 lipid binding chaperone B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080533.2 233 Missense Mutation ATG,CTG M65L NP_001074002.1

View Full Product Details