Product Details

SNP ID
rs9684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87196306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTATATTCAGTTTAACAGAAA[A/T]AAAAGAATATTTGTCTTAAGATGCA
Phenotype
MIM: 608491 MIM: 616993
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DMTF1 PubMed Links

Gene Details

Gene
DMTF1
Gene Name
cyclin D binding myb like transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142326.1 982 UTR 3 NP_001135798.1
NM_001142327.1 982 UTR 3 NP_001135799.1
NM_021145.3 982 UTR 3 NP_066968.3
XM_011516735.2 982 UTR 3 XP_011515037.1
XM_011516737.1 982 UTR 3 XP_011515039.1
XM_011516739.2 982 UTR 3 XP_011515041.1
XM_017012865.1 982 UTR 3 XP_016868354.1
XM_017012866.1 982 UTR 3 XP_016868355.1
XM_017012867.1 982 UTR 3 XP_016868356.1
XM_017012868.1 982 UTR 3 XP_016868357.1
XM_017012869.1 982 UTR 3 XP_016868358.1
XM_017012870.1 982 UTR 3 XP_016868359.1
XM_017012871.1 982 UTR 3 XP_016868360.1
XM_017012872.1 982 UTR 3 XP_016868361.1
XM_017012873.1 982 UTR 3 XP_016868362.1
XM_017012874.1 982 UTR 3 XP_016868363.1
XM_017012875.1 982 UTR 3 XP_016868364.1
XM_017012876.1 982 UTR 3 XP_016868365.1
XM_017012877.1 982 UTR 3 XP_016868366.1
XM_017012878.1 982 UTR 3 XP_016868367.1
Gene
TMEM243
Gene Name
transmembrane protein 243
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005250586.3 982 Intron XP_005250643.1
XM_005250587.4 982 Intron XP_005250644.1
XM_005250588.3 982 Intron XP_005250645.1
XM_005250589.3 982 UTR 3 XP_005250646.1
XM_017012616.1 982 Intron XP_016868105.1
XM_017012617.1 982 UTR 3 XP_016868106.1
XM_017012618.1 982 UTR 3 XP_016868107.1

View Full Product Details