Product Details

SNP ID
rs8109823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32692410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAGGACTTCCCCACCAGGCCC[A/G]GCACCCGGGCCTGGCCTCGCCCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT19 PubMed Links

Gene Details

Gene
NUDT19
Gene Name
nudix hydrolase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105570.1 804 Silent Mutation CCA,CCG P150P NP_001099040.1
XM_017026806.1 804 Silent Mutation CCA,CCG P150P XP_016882295.1

View Full Product Details