Product Details

SNP ID
rs1957374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22766599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCGTTCTCTTTTACCGCCTGATCT[A/G]CGGCTACAATTCTGGCTTCATGCCC
Phenotype
MIM: 601066 MIM: 603593
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OXA1L PubMed Links

Gene Details

Gene
OXA1L
Gene Name
OXA1L, mitochondrial inner membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005015.3 78 Silent Mutation CTA,CTG L26L NP_005006.3
Gene
SLC7A7
Gene Name
solute carrier family 7 member 7
There are no transcripts associated with this gene.

View Full Product Details