Product Details

SNP ID
rs4674679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222682705 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTGTCAACAGATGCATGAATTA[G/T]CAGATTACAGTGTATATGTAAGTGG
Phenotype
MIM: 610268
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MOGAT1 PubMed Links

Gene Details

Gene
MOGAT1
Gene Name
monoacylglycerol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058165.2 Intron NP_477513.2

View Full Product Details