Product Details

SNP ID
rs4292120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240126863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGCATCTGGTAAACATTAGCGC[C/T]TCCGCCCCCTGCCCATGGCCTGTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPS9 PubMed Links

Gene Details

Gene
COPS9
Gene Name
COP9 signalosome subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163424.1 884 Intron NP_001156896.1
NM_138336.1 884 Silent Mutation GAA,GAG E153E NP_612209.1
XM_017003411.1 884 Silent Mutation GAA,GAG E143E XP_016858900.1

View Full Product Details