Product Details

SNP ID
rs17567
Assay Type
Validated
NCBI dbSNP Submissions
89
Location
Chr.1:51361249 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGCAGAAGTGAATGGATCACAAAA[C/T]ATTTCAGGATCTTTTTCTTTGGGGC
Phenotype
MIM: 600051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPS15 PubMed Links

Gene Details

Gene
EPS15
Gene Name
epidermal growth factor receptor pathway substrate 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159969.1 2499 Missense Mutation ATA,ATG I508M NP_001153441.1
NM_001981.2 2499 Missense Mutation ATA,ATG I822M NP_001972.1
XM_005270618.2 2499 Missense Mutation ATA,ATG I792M XP_005270675.1
XM_017000615.1 2499 Missense Mutation ATA,ATG I859M XP_016856104.1
XM_017000616.1 2499 Missense Mutation ATA,ATG I791M XP_016856105.1
XM_017000617.1 2499 Missense Mutation ATA,ATG I754M XP_016856106.1
XM_017000618.1 2499 Missense Mutation ATA,ATG I724M XP_016856107.1
XM_017000619.1 2499 Missense Mutation ATA,ATG I634M XP_016856108.1
XM_017000620.1 2499 Missense Mutation ATA,ATG I584M XP_016856109.1

View Full Product Details