Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001185118.1 | 1205 | Silent Mutation | CAC,CAT | H6H | NP_001172047.1 |
NM_001185119.1 | 1205 | Silent Mutation | CAC,CAT | H6H | NP_001172048.1 |
NM_018657.4 | 1205 | Silent Mutation | CAC,CAT | H6H | NP_061127.1 |
XM_005247621.4 | 1205 | Silent Mutation | CAC,CAT | H6H | XP_005247678.1 |
XM_005247622.3 | 1205 | Silent Mutation | CAC,CAT | H6H | XP_005247679.1 |
XM_005247624.3 | 1205 | UTR 5 | XP_005247681.1 | ||
XM_011512987.1 | 1205 | Silent Mutation | CAC,CAT | H6H | XP_011511289.1 |
XM_017006864.1 | 1205 | Silent Mutation | CAC,CAT | H6H | XP_016862353.1 |
XM_017006865.1 | 1205 | UTR 5 | XP_016862354.1 | ||
XM_017006866.1 | 1205 | UTR 5 | XP_016862355.1 | ||
XM_017006867.1 | 1205 | UTR 5 | XP_016862356.1 | ||
XM_017006868.1 | 1205 | UTR 5 | XP_016862357.1 |