Product Details

SNP ID
rs2041969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20389246 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGAAGGAGGACGAATGAAGAAATG[A/C]AAACCAAGGAAAAAAGGAGCTGTTT
Phenotype
MIM: 194548
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
USP41 PubMed Links

Gene Details

Gene
USP41
Gene Name
ubiquitin specific peptidase 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017029164.1 Intron XP_016884653.1
Gene
ZNF74
Gene Name
zinc finger protein 74
There are no transcripts associated with this gene.

View Full Product Details