Product Details

SNP ID
rs7589786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:11156294 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCAGTTGGGAAGGGGTGACGGA[A/G]AGGTCGTTAAATCCTTGATTCTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf50 PubMed Links

Gene Details

Gene
C2orf50
Gene Name
chromosome 2 open reading frame 50
There are no transcripts associated with this gene.

Gene
PQLC3
Gene Name
PQ loop repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282710.1 Intron NP_001269639.1
NM_001282711.1 Intron NP_001269640.1
NM_001282712.1 Intron NP_001269641.1
NM_152391.4 Intron NP_689604.1
XM_011510315.1 Intron XP_011508617.1

View Full Product Details