Product Details

SNP ID
rs3764885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23783476 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTGGAGGTGGCTCCGTTTCCCAG[A/G]GTGGGGTGATTCACGTCTTGAGGTC
Phenotype
MIM: 313020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAT1 PubMed Links

Gene Details

Gene
SAT1
Gene Name
spermidine/spermine N1-acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002970.3 Intron NP_002961.1
XM_017029720.1 Intron XP_016885209.1

View Full Product Details