Product Details

SNP ID
rs6642730
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:110015501 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCTTCATATCAGCACTTGTTCATT[A/G]TTGGTTTGCATTTGTTAAGAGTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM164 PubMed Links

Gene Details

Gene
TMEM164
Gene Name
transmembrane protein 164
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017698.2 4559 Intron NP_060168.2
NM_032227.3 4559 Intron NP_115603.2
XM_005262205.4 4559 Intron XP_005262262.1
XM_005262206.3 4559 Intron XP_005262263.1
XM_005262208.4 4559 Intron XP_005262265.1
XM_011531051.2 4559 Intron XP_011529353.1
XM_011531054.1 4559 Intron XP_011529356.1
XM_011531055.2 4559 Intron XP_011529357.1
XM_017029894.1 4559 Intron XP_016885383.1
XM_017029895.1 4559 UTR 3 XP_016885384.1
XM_017029896.1 4559 Intron XP_016885385.1
XM_017029897.1 4559 Intron XP_016885386.1
XM_017029898.1 4559 Intron XP_016885387.1
XM_017029899.1 4559 Intron XP_016885388.1
XM_017029900.1 4559 Intron XP_016885389.1

View Full Product Details