Product Details

SNP ID
rs28684959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43849810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCATAGCTGGGACCTATCCCAA[A/T]CCCAGGCCCATCCCTACCACCATAG
Phenotype
MIM: 610520
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD300LG PubMed Links

Gene Details

Gene
CD300LG
Gene Name
CD300 molecule like family member g
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168322.1 Intron NP_001161794.1
NM_001168323.1 Intron NP_001161795.1
NM_001168324.1 Intron NP_001161796.1
NM_145273.3 Intron NP_660316.2
XM_005257075.2 Intron XP_005257132.1
XM_005257076.2 Intron XP_005257133.1
XM_006721721.2 Intron XP_006721784.1
XM_006721722.2 Intron XP_006721785.1
XM_011524384.2 Intron XP_011522686.1
XM_017024235.1 Intron XP_016879724.1

View Full Product Details