Product Details

SNP ID
rs2071452
Assay Type
Validated
NCBI dbSNP Submissions
24
Location
Chr.1:203083721 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
CTCTAAGGAGGCAGCTGAATGAGGG[C/T]GTCCAGTCCCTTGCTGGGGGGTGGG
Phenotype
MIM: 159980 MIM: 603145
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYOG PubMed Links

Gene Details

Gene
MYOG
Gene Name
myogenin (myogenic factor 4)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002479.5 941 UTR 3 NP_002470.2
Gene
PPFIA4
Gene Name
PTPRF interacting protein alpha 4
There are no transcripts associated with this gene.

View Full Product Details