Product Details
- SNP ID
-
rs1953325
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:52236688 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACAAACACCTGAAGTATTTTTAA[A/C]TTATTAATACTTTATTATATTAGCA
- Phenotype
-
MIM: 606496
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL17F
PubMed Links
Gene Details
- Gene
- IL17F
- Gene Name
- interleukin 17F
View Full Product Details