Product Details
- SNP ID
-
hCV11915213
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:46856610 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTTTGAGGATGCAATATATTTAG[A/C]CTAATTGATCAAAATCCCAAAGAAA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRF5
PubMed Links
Gene Details
- Gene
- ADGRF5
- Gene Name
- adhesion G protein-coupled receptor F5
View Full Product Details