Product Details

SNP ID
rs1131865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219289814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCAGAGGCAGGCCCGGGGCTGA[A/G]AAGCAGGGGGAGCCGGGTGTGGCGA
Phenotype
MIM: 604139 MIM: 601773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB2 PubMed Links

Gene Details

Gene
DNAJB2
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
There are no transcripts associated with this gene.

Gene
MIR153-1
Gene Name
microRNA 153-1
There are no transcripts associated with this gene.

Gene
PTPRN
Gene Name
protein tyrosine phosphatase, receptor type N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199763.1 3136 UTR 3 NP_001186692.1
NM_001199764.1 3136 UTR 3 NP_001186693.1
NM_002846.3 3136 UTR 3 NP_002837.1
XM_011511566.1 3136 UTR 3 XP_011509868.1
XM_017004609.1 3136 UTR 3 XP_016860098.1
XM_017004610.1 3136 UTR 3 XP_016860099.1
XM_017004611.1 3136 UTR 3 XP_016860100.1
XM_017004612.1 3136 UTR 3 XP_016860101.1

View Full Product Details