Product Details

SNP ID
rs10139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:229767539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGCTGTAACAGGCAGACACTG[C/T]ATTTCTTGCTATGATCAGGAAAGAT
Phenotype
MIM: 604506
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIP12 PubMed Links

Gene Details

Gene
TRIP12
Gene Name
thyroid hormone receptor interactor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284214.1 6262 UTR 3 NP_001271143.1
NM_001284215.1 6262 UTR 3 NP_001271144.1
NM_001284216.1 6262 UTR 3 NP_001271145.1
NM_004238.2 6262 UTR 3 NP_004229.1
XM_005246954.4 6262 UTR 3 XP_005247011.1
XM_005246955.4 6262 UTR 3 XP_005247012.1
XM_005246956.4 6262 UTR 3 XP_005247013.1
XM_005246957.4 6262 UTR 3 XP_005247014.1
XM_005246958.4 6262 UTR 3 XP_005247015.1
XM_005246960.4 6262 UTR 3 XP_005247017.1
XM_005246961.3 6262 UTR 3 XP_005247018.1
XM_005246962.4 6262 UTR 3 XP_005247019.1
XM_006712852.3 6262 UTR 3 XP_006712915.1
XM_011512180.2 6262 UTR 3 XP_011510482.1
XM_011512181.2 6262 UTR 3 XP_011510483.1
XM_011512183.2 6262 UTR 3 XP_011510485.1
XM_017005272.1 6262 UTR 3 XP_016860761.1
XM_017005273.1 6262 UTR 3 XP_016860762.1
XM_017005274.1 6262 UTR 3 XP_016860763.1
XM_017005275.1 6262 UTR 3 XP_016860764.1
XM_017005276.1 6262 UTR 3 XP_016860765.1
XM_017005277.1 6262 UTR 3 XP_016860766.1
XM_017005278.1 6262 UTR 3 XP_016860767.1
XM_017005279.1 6262 UTR 3 XP_016860768.1
XM_017005280.1 6262 UTR 3 XP_016860769.1
XM_017005281.1 6262 UTR 3 XP_016860770.1
XM_017005282.1 6262 UTR 3 XP_016860771.1
XM_017005283.1 6262 UTR 3 XP_016860772.1
XM_017005284.1 6262 UTR 3 XP_016860773.1
XM_017005285.1 6262 UTR 3 XP_016860774.1
XM_017005286.1 6262 UTR 3 XP_016860775.1
XM_017005287.1 6262 UTR 3 XP_016860776.1
XM_017005288.1 6262 UTR 3 XP_016860777.1
XM_017005289.1 6262 UTR 3 XP_016860778.1
XM_017005290.1 6262 UTR 3 XP_016860779.1
XM_017005291.1 6262 UTR 3 XP_016860780.1
XM_017005292.1 6262 UTR 3 XP_016860781.1
XM_017005293.1 6262 UTR 3 XP_016860782.1

View Full Product Details