Product Details
- SNP ID
-
rs15413
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:85342256 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCTAACCCTCACATGTGCTAATTAA[C/T]TGCAAATGCCATTTCTGGGCTTCAC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RETSAT
PubMed Links
Gene Details
- Gene
- RETSAT
- Gene Name
- retinol saturase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017750.3 |
3010 |
UTR 3 |
|
|
NP_060220.3 |
View Full Product Details