Product Details

SNP ID
rs2047091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100604040 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTCTTACACCTCAGATTTTCTAG[A/G]AGAACTTTTAAAAATTTGCACAATA
Phenotype
MIM: 615052 MIM: 610350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB7 PubMed Links

Gene Details

Gene
ASB7
Gene Name
ankyrin repeat and SOCS box containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024708.3 Intron NP_078984.2
NM_198243.2 Intron NP_937886.1
Gene
LINS1
Gene Name
lines homolog 1
There are no transcripts associated with this gene.

View Full Product Details