Product Details
- SNP ID
-
hCV11949755
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88775329 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATTGGATTATTGTTGCTTCAGAG[A/C]TTACTGGAAGCAGAAGGTGGAAGAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD36BP2
PubMed Links
Gene Details
- Gene
- ANKRD36BP2
- Gene Name
- ankyrin repeat domain 36B pseudogene 2
There are no transcripts associated with this gene.
View Full Product Details