Product Details

SNP ID
rs1991659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:88060883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGAGTTGACATTTTTCAGGTAA[A/G]TTTTTAATGCTTTAATTTAAATTGC
Phenotype
MIM: 603363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CGGBP1 PubMed Links

Gene Details

Gene
CGGBP1
Gene Name
CGG triplet repeat binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008390.1 Intron NP_001008391.1
NM_001195308.1 Intron NP_001182237.1
NM_003663.3 Intron NP_003654.3
XM_017007355.1 Intron XP_016862844.1
Gene
ZNF654
Gene Name
zinc finger protein 654
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018293.2 Intron NP_060763.2
XM_011534340.2 Intron XP_011532642.2
XM_017006789.1 Intron XP_016862278.1
XM_017006790.1 Intron XP_016862279.1
XM_017006791.1 Intron XP_016862280.1
XM_017006792.1 Intron XP_016862281.1
XM_017006793.1 Intron XP_016862282.1
XM_017006794.1 Intron XP_016862283.1
XM_017006795.1 Intron XP_016862284.1

View Full Product Details