Product Details

SNP ID
rs14791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78991766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAAAACAGATGGATCCCATAGG[A/G]AAGGAACACAATCATTATGCTGACA
Phenotype
MIM: 613165
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CANT1 PubMed Links

Gene Details

Gene
CANT1
Gene Name
calcium activated nucleotidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159772.1 3408 UTR 3 NP_001153244.1
NM_001159773.1 3408 UTR 3 NP_001153245.1
NM_138793.3 3408 UTR 3 NP_620148.1
XM_005257021.1 3408 UTR 3 XP_005257078.1
XM_005257022.1 3408 UTR 3 XP_005257079.1
XM_006721683.1 3408 UTR 3 XP_006721746.1
XM_011524291.1 3408 UTR 3 XP_011522593.1
XM_011524293.1 3408 UTR 3 XP_011522595.1
XM_011524294.1 3408 UTR 3 XP_011522596.1
XM_011524295.2 3408 UTR 3 XP_011522597.1

View Full Product Details