Product Details

SNP ID
rs2054987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:56559077 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCCTTCTAGCCAAATAAAATCA[C/G]ATTACAACACTGCTTACTACTGTCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC66 PubMed Links

Gene Details

Gene
CCDC66
Gene Name
coiled-coil domain containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012506.4 Intron NP_001012524.4
NM_001141947.1 Intron NP_001135419.1
XM_005265081.3 Intron XP_005265138.1
XM_005265082.3 Intron XP_005265139.1
XM_005265083.3 Intron XP_005265140.1
XM_005265084.3 Intron XP_005265141.1
XM_011533614.2 Intron XP_011531916.1
XM_011533615.2 Intron XP_011531917.1
XM_011533616.2 Intron XP_011531918.1
XM_011533617.2 Intron XP_011531919.1
XM_011533619.2 Intron XP_011531921.1
XM_017006229.1 Intron XP_016861718.1
XM_017006230.1 Intron XP_016861719.1
XM_017006231.1 Intron XP_016861720.1
XM_017006232.1 Intron XP_016861721.1
XM_017006233.1 Intron XP_016861722.1
XM_017006234.1 Intron XP_016861723.1
XM_017006235.1 Intron XP_016861724.1
XM_017006236.1 Intron XP_016861725.1
XM_017006237.1 Intron XP_016861726.1
XM_017006238.1 Intron XP_016861727.1
XM_017006239.1 Intron XP_016861728.1
XM_017006240.1 Intron XP_016861729.1
XM_017006241.1 Intron XP_016861730.1
XM_017006242.1 Intron XP_016861731.1
XM_017006243.1 Intron XP_016861732.1
XM_017006244.1 Intron XP_016861733.1
XM_017006245.1 Intron XP_016861734.1

View Full Product Details