Product Details

SNP ID
rs9976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45306542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGGAAAAGAAGAGGACCCTGC[C/T]AGGGAGATATTTCATTGGCCAGAAT
Phenotype
MIM: 123310 MIM: 606495
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CKM PubMed Links

Gene Details

Gene
CKM
Gene Name
creatine kinase, M-type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001824.4 1529 UTR 3 NP_001815.2
XM_017026240.1 1529 UTR 3 XP_016881729.1
Gene
MARK4
Gene Name
microtubule affinity regulating kinase 4
There are no transcripts associated with this gene.

View Full Product Details