Product Details

SNP ID
rs3501
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:154946699 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCACCGAGTTCTGCTTGTGCTCCA[T/C]GAGCTTCCGGAAAGATGGCTGCTTG
Phenotype
MIM: 608254
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
PAXIP1 PubMed Links

Gene Details

Gene
PAXIP1
Gene Name
PAX interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007349.3 4392 Missense Mutation NP_031375.3
XM_005249539.1 4392 Missense Mutation XP_005249596.1
XM_011515982.2 4392 Missense Mutation XP_011514284.1
XM_011515983.1 4392 Missense Mutation XP_011514285.1
XM_017011881.1 4392 Missense Mutation XP_016867370.1
Gene
PAXIP1-AS2
Gene Name
PAXIP1 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details