Product Details

SNP ID
rs364734
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:73599678 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTAAGTGAGAGGAAATAGTAACAG[G/T]TACCGTGTCACTTAAGGAATTAGAA
Phenotype
MIM: 604322
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC17A5 PubMed Links

Gene Details

Gene
SLC17A5
Gene Name
solute carrier family 17 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012434.4 Intron NP_036566.1
XM_005248710.3 Intron XP_005248767.1
XM_005248711.2 Intron XP_005248768.1
XM_011535750.2 Intron XP_011534052.1
XM_017010730.1 Intron XP_016866219.1
XM_017010731.1 Intron XP_016866220.1

View Full Product Details