Product Details

SNP ID
rs1023070454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37990804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTAGCACTGCCCACAGGCAGGGC[A/G]AAGGCGCGGGAGCAGTTTCCAGCAG
Phenotype
MIM: 608592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTDSPL PubMed Links

Gene Details

Gene
CTDSPL
Gene Name
CTD small phosphatase like
There are no transcripts associated with this gene.

Gene
VILL
Gene Name
villin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015873.3 Intron NP_056957.3

View Full Product Details