Product Details

SNP ID
rs1929095
Assay Type
Validated
NCBI dbSNP Submissions
40
Location
Chr.1:186443589 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGACTTCAAATAGAAAATTTATTT[C/T]CTCATGAGATTTCATTTTTATTACA
Phenotype
MIM: 171490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724919 PubMed Links

Gene Details

Gene
LOC102724919
Gene Name
uncharacterized LOC102724919
There are no transcripts associated with this gene.

Gene
PDC
Gene Name
phosducin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002597.4 1205 UTR 3 NP_002588.3
NM_022576.3 1205 UTR 3 NP_072098.1
XM_011509603.2 1205 UTR 3 XP_011507905.1

View Full Product Details