Product Details

SNP ID
rs1854797
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:92833199 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CATTTTTATTGTTTTATGAAACTAC[C/T]AGTCTGTGACATGGAAGGTAGAGGA
Phenotype
MIM: 614542 MIM: 603634 MIM: 603635
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM69A PubMed Links

Gene Details

Gene
FAM69A
Gene Name
family with sequence similarity 69 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006605.4 Intron NP_001006606.2
NM_001252269.1 Intron NP_001239198.1
NM_001252270.1 Intron NP_001239199.1
NM_001252271.1 Intron NP_001239200.1
NM_001252273.1 Intron NP_001239202.1
Gene
RPL5
Gene Name
ribosomal protein L5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000969.3 Intron NP_000960.2
Gene
SNORA66
Gene Name
small nucleolar RNA, H/ACA box 66
There are no transcripts associated with this gene.

Gene
SNORD21
Gene Name
small nucleolar RNA, C/D box 21
There are no transcripts associated with this gene.

View Full Product Details