Product Details

SNP ID
rs1843221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88773861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTATATATTTCAGATCTATTCAG[G/T]TTGACACAATTTGGTCATCTTTGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD36BP2 PubMed Links

Gene Details

Gene
ANKRD36BP2
Gene Name
ankyrin repeat domain 36B pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details