Product Details
- SNP ID
-
rs1819318
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:22209528 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGGAGGCTGTTGGTGCATATCGG[G/A]TCACTCGATCCGGCAGCAATGCCTA
- Phenotype
-
MIM: 613249
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EBLN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10466081] are located under a probe and SNP(s) [rs3059425] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EBLN1
- Gene Name
- endogenous Bornavirus-like nucleoprotein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199938.1 |
456 |
Silent Mutation |
GAC,GAT |
D152D |
NP_001186867.1 |
View Full Product Details