Product Details

SNP ID
hCV12015834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:88294058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCTATGCTCTCAATTGCACAC[C/T]CATCAGTTCCTCTGCGGATCCGGAA
Phenotype
MIM: 602365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSC PubMed Links

Gene Details

Gene
CTSC
Gene Name
cathepsin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114173.2 1455 Intron NP_001107645.1
NM_001814.5 1455 Missense Mutation GAG,GGG E447G NP_001805.3
NM_148170.4 1455 Intron NP_680475.1

View Full Product Details