Product Details
- SNP ID
-
rs14254
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:135933281 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGAGAGCAGGAGCAGCTGCAGCA[C/T]CTCTAACAGTCCAGGGCTGAGGCGC
- Phenotype
-
MIM: 608129
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBAC1
PubMed Links
Gene Details
- Gene
- UBAC1
- Gene Name
- UBA domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016172.2 |
1555 |
UTR 3 |
|
|
NP_057256.2 |
View Full Product Details