Product Details
- SNP ID
-
rs4698
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:10307220 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTCCTCCCCTATTTTAAGACGTGA[A/G]TGTCTCAGCGAGGTGTAAAGTTGTT
- Phenotype
-
MIM: 103275
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34354539] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADM
- Gene Name
- adrenomedullin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001124.2 |
1408 |
UTR 3 |
|
|
NP_001115.1 |
- Gene
- CAND1.11
- Gene Name
- uncharacterized LOC100130460
There are no transcripts associated with this gene.
- Gene
- LOC105376544
- Gene Name
- uncharacterized LOC105376544
There are no transcripts associated with this gene.
View Full Product Details