Product Details

SNP ID
rs1057227
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41770420 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTCGCCTGGTCACCCTTCTCT[C/T]CCCAGCCTGGAGACAGACCAGGAGA
Phenotype
MIM: 607744
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FRS3 PubMed Links

Gene Details

Gene
FRS3
Gene Name
fibroblast growth factor receptor substrate 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006653.4 2788 UTR 3 NP_006644.1
XM_011514254.2 2788 UTR 3 XP_011512556.1
XM_011514255.2 2788 UTR 3 XP_011512557.1
XM_017010193.1 2788 UTR 3 XP_016865682.1

View Full Product Details