Product Details

SNP ID
rs6274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113410206 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGCTTGGGAATCTCTGGGGTCCA[A/C]CCTGCAGTCTGGTATTTACATGGGT
Phenotype
MIM: 608774 MIM: 126450
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKK1 PubMed Links

Gene Details

Gene
ANKK1
Gene Name
ankyrin repeat and kinase domain containing 1
There are no transcripts associated with this gene.

Gene
DRD2
Gene Name
dopamine receptor D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000795.3 2070 UTR 3 NP_000786.1
NM_016574.3 2070 UTR 3 NP_057658.2
XM_017017296.1 2070 UTR 3 XP_016872785.1

View Full Product Details