Product Details

SNP ID
rs6241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27958161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTTATTTTTACATGCACAAAGG[A/G]AAATCAATTTGGATAATTATATTTA
Phenotype
MIM: 168470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTHLH PubMed Links

Gene Details

Gene
PTHLH
Gene Name
parathyroid hormone like hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002820.2 3790 Intron NP_002811.1
NM_198964.1 3790 Intron NP_945315.1
NM_198965.1 3790 UTR 3 NP_945316.1
NM_198966.1 3790 UTR 3 NP_945317.1
XM_011520774.2 3790 UTR 3 XP_011519076.1
XM_011520775.2 3790 UTR 3 XP_011519077.1
XM_017019674.1 3790 UTR 3 XP_016875163.1
XM_017019675.1 3790 UTR 3 XP_016875164.1

View Full Product Details