Product Details

SNP ID
rs967
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27961710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTCATTCACTGTGCTCAAGATT[A/T]TTCTAGTGCCACTGCCCATTGATTA
Phenotype
MIM: 168470
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
PTHLH PubMed Links

Gene Details

Gene
PTHLH
Gene Name
parathyroid hormone like hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002820.2 Intron NP_002811.1
NM_198964.1 Intron NP_945315.1
NM_198965.1 Intron NP_945316.1
NM_198966.1 Intron NP_945317.1
XM_011520774.2 Intron XP_011519076.1
XM_011520775.2 Intron XP_011519077.1
XM_017019674.1 Intron XP_016875163.1
XM_017019675.1 Intron XP_016875164.1

View Full Product Details