Product Details
- SNP ID
-
rs2923
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:127706044 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAAATTTGTGGAGATGCCCCATGT[C/G]TTGTGAGACTTAAAAAAAAGAAAAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SFT2D3
PubMed Links
Gene Details
- Gene
- SFT2D3
- Gene Name
- SFT2 domain containing 3
There are no transcripts associated with this gene.
- Gene
- WDR33
- Gene Name
- WD repeat domain 33
View Full Product Details