Product Details

SNP ID
rs11516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53064321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGTAGGATCACATGAATAGGG[A/G]AAAAAAGAGAGTCTATTTTTGTCTA
Phenotype
MIM: 607717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC283335 PubMed Links

Gene Details

Gene
LOC283335
Gene Name
uncharacterized LOC283335
There are no transcripts associated with this gene.

Gene
MIR6757
Gene Name
microRNA 6757
There are no transcripts associated with this gene.

Gene
SPRYD3
Gene Name
SPRY domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032840.2 4722 UTR 3 NP_116229.1
Gene
TNS2
Gene Name
tensin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015319.2 4722 UTR 3 NP_056134.2
NM_170754.2 4722 UTR 3 NP_736610.2
NM_198316.1 4722 UTR 3 NP_938072.1
XM_006719302.3 4722 UTR 3 XP_006719365.1
XM_006719303.1 4722 UTR 3 XP_006719366.1
XM_011538079.1 4722 UTR 3 XP_011536381.1
XM_017019088.1 4722 UTR 3 XP_016874577.1
XM_017019089.1 4722 UTR 3 XP_016874578.1

View Full Product Details