Product Details
- SNP ID
-
rs2020
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:53065813 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGAGGGTGAGCAGGGAGAAGGAG[A/C]AGGTCTGGAGGGGAGGCCCTAGCCA
- Phenotype
-
MIM: 607717
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6757
PubMed Links
Gene Details
- Gene
- MIR6757
- Gene Name
- microRNA 6757
There are no transcripts associated with this gene.
- Gene
- SPRYD3
- Gene Name
- SPRY domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032840.2 |
1435 |
UTR 3 |
|
|
NP_116229.1 |
- Gene
- TNS2
- Gene Name
- tensin 2
There are no transcripts associated with this gene.
View Full Product Details