Product Details

SNP ID
rs14920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67657612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGTCCTGCGTGACGTCTCACAT[A/C]GGAGTTGGCTCAGACCCTGGCTGTG
Phenotype
MIM: 609377 MIM: 610859 MIM: 607484
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACD PubMed Links

Gene Details

Gene
ACD
Gene Name
adrenocortical dysplasia homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082486.1 1966 Silent Mutation NP_001075955.1
NM_001082487.1 1966 Silent Mutation NP_001075956.1
NM_022914.2 1966 Silent Mutation NP_075065.2
XM_005256115.3 1966 Silent Mutation XP_005256172.1
Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
CARMIL2
Gene Name
capping protein regulator and myosin 1 linker 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013838.1 1966 Intron NP_001013860.1
NM_001317026.1 1966 Intron NP_001303955.1
XM_011522874.1 1966 Intron XP_011521176.1
XM_011522875.2 1966 Intron XP_011521177.2
XM_017022953.1 1966 Intron XP_016878442.1
Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
PARD6A
Gene Name
par-6 family cell polarity regulator alpha
There are no transcripts associated with this gene.

View Full Product Details