Product Details
- SNP ID
-
hCV12075047
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:82811330 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTTCAACTTTATCGTCTTCACCT[T/G]AGGATATGAGGAACCCACAGGGGAG
- Phenotype
-
MIM: 612393
- Polymorphism
- T/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FSD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142681702,rs145800406] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FSD2
- Gene Name
- fibronectin type III and SPRY domain containing 2
There are no transcripts associated with this gene.
- Gene
- WHAMM
- Gene Name
- WAS protein homolog associated with actin, golgi membranes and microtubules
View Full Product Details