Product Details

SNP ID
rs1687257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184252007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACAAGTAAACACTTTCTTGAAT[C/G]TGTGATTTTAAACCATTGCACTTAT
Phenotype
MIM: 608750 MIM: 608721 MIM: 610145
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALG3 PubMed Links

Gene Details

Gene
ALG3
Gene Name
ALG3, alpha-1,3- mannosyltransferase
There are no transcripts associated with this gene.

Gene
CAMK2N2
Gene Name
calcium/calmodulin dependent protein kinase II inhibitor 2
There are no transcripts associated with this gene.

Gene
ECE2
Gene Name
endothelin converting enzyme 2
There are no transcripts associated with this gene.

Gene
VWA5B2
Gene Name
von Willebrand factor A domain containing 5B2
There are no transcripts associated with this gene.

View Full Product Details