Product Details

SNP ID
rs1800584
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:18130781 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
AGACAACGTATATTGCATATTTTAC[C/T]TGAAACAAGAAAGAGTAACATGTTA
Phenotype
MIM: 608072 MIM: 187680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NHLRC1 PubMed Links

Gene Details

Gene
NHLRC1
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

Gene
TPMT
Gene Name
thiopurine S-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000367.3 Intron NP_000358.1
XM_011514839.2 Intron XP_011513141.1
XM_017011241.1 Intron XP_016866730.1

View Full Product Details