Product Details

SNP ID
rs10848723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2895854 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCTCCTAGGCTACAAAGCTATAC[A/G]GCCTGTTACTGTACTGAATATTGTA
Phenotype
MIM: 614085 MIM: 604730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHNO1 PubMed Links

Gene Details

Gene
RHNO1
Gene Name
RAD9-HUS1-RAD1 interacting nuclear orphan 1
There are no transcripts associated with this gene.

Gene
TULP3
Gene Name
tubby like protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160408.1 Intron NP_001153880.1
NM_003324.4 Intron NP_003315.2

View Full Product Details